Strontium oxide, 99.5% (metals basis), SrO »97%, Alfa Aesar™

Manufacturer: Alfa Aesar 8822006

Quantity 5g

 View more versions of this product

Catalog No. AA8822006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-11-0
    Physical Form -100 Mesh Powder
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula SrO
    Formula Weight 103.62
    MDL Number MFCD00049558
    Chemical Name or Material Strontium oxide

    Specifications

    Physical Form -100 Mesh Powder
    Formula Weight 103.62
    Quantity 5g
    EINECS Number 215-219-9
    Density 4.7g/mL
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula SrO
    MDL Number MFCD00049558
    CAS 1314-11-0
    Chemical Name or Material Strontium oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit