Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Strontium ranelate

Catalog No. AC463010250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463010250 25 g
AC463010050 5 g
AC463011000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC463010250 Supplier Thermo Scientific Chemicals Supplier No. 463010250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135459-87-9
Molecular Formula C12H6N2O8SSr2

Specifications

Quantity 25 g
Formula Weight 513.49
InChI Key KVJIIORUFXGTFN-UHFFFAOYSA-L
SMILES [Sr++].OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O.OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O
IUPAC Name strontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate)
Molecular Weight (g/mol) 770.16
Chemical Name or Material Strontium ranelate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.