Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium sulfide, 99.9% (metals basis)

Catalog No. p-7080533
Click to view available options
Quantity:
10 g
50 g

CAS: 1314-96-1 | SSr | 119.68 g/mol

Strontium sulfide is used in chemical research and as organic intermediates. It is also used as depilatory. It is also used as Isotope labelled enantiomer of Voriconazole (V760000), an antifungal agent and an ergosterol biosynthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-96-1
Molecular Formula SSr
Molecular Weight (g/mol) 119.68
MDL Number MFCD00049560
InChI Key ZEGFMFQPWDMMEP-UHFFFAOYSA-N
Synonym strontium sulfide, strontium monosulfide, strontium sulphide, strontiumsulfid, hsdb 12
PubChem CID 14820
IUPAC Name strontium(2+) sulfanediide
SMILES [S--].[Sr++]

Specifications

Melting Point >2000°C
Color Gray
Physical Form Powder
Quantity 10 g
Merck Index 14,8849
Solubility Information Slightly soluble in water; Decomposes in acids
Formula Weight 119.68
Percent Purity 99.9%
Odor Like rotten eggs
Sensitivity Hygroscopic
Density 3.70 g/mL
Assay (metals basis)
Chemical Name or Material Strontium sulfide
Show More Show Less
Hazard Category H290-H301-H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
EINECSNumber 215-249-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.