Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Structure of the Sun 3D Model Making Kit
SCI_ED

Catalog No. S436051 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436051 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436051 Supplier NewPath Learning™ Supplier No. 247818
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Structure of the Sun 3D Model Making Kit allows you to teach about the exterior and interior structure of our Sun!

  • Your students begin by making their model of the Sun, and then they use it to learn about the structure of the Sun.
  • First they look at the exterior features of the Sun.
    • This includes the Sun's atmosphere (corona, chromosphere), the Sun's surface (photosphere), and active phenomena such as sun spots, solar prominences and solar flares.
  • Next they look at the interior structure of the Sun and what each layer does.
    • This covers the core, radiation (radiative) zone, tachocline, and convection (convective) zone.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Product Type 3D Model
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Width (English) 9 in.
Height (English) 12 in.
Depth (English) 2 in.
Material Paper
Languages English
For Use With (Application) Model Assembly
Subject STEM, Astronomy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.