Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sts-1Rabbit anti-Mouse, Polyclonal, Rockland™
SDP

Catalog No. 501991826
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1826 Supplier Rockland Immunochemicals Supplier No. 600401870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Sts-1 protein. A BLAST analysis was used to suggest reactivity with this protein from mouse, human, rat and dog based on 100% homology for the immunogen sequence. Cross reactivity with Sts-1 homologues from other sources has not been determined.

Sts1 encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor.

Specifications

Antigen Sts-1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene UBASH3B
Gene Accession No. Q8BGG7
Gene Alias 2810457I06Rik; BB125008; BOS_14514; Cbl-interacting protein p70; Cbl-interacting protein Sts-1; KIAA1959; LOC100337868; MGC15437; nm23-phosphorylated unknown substrate; p70; RGD1310357; SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling 1, nm23-phosphorylated unknown substrate; STS1; STS-1; Suppressor of T-cell receptor signaling 1; T-cell ubiquitin ligand 2; TULA2; TULA-2; Tyrosine-protein phosphatase STS1/TULA2; UBASH3B; ubiquitin associated and SH3 domain containing B; ubiquitin associated and SH3 domain containing, B; Ubiquitin-associated and SH3 domain-containing protein B
Gene Symbols UBASH3B
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72828
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.