Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542055
Description
Peptide sequence: EPTSNMDIFP TVAKLAGAPL PEDRIIDGRD LMPLLEGKSQ RSDHEFLFHY Sequence homology: Cow: 90%; Dog: 80%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 77%; Pig: 92%; Rabbit: 77%; Zebrafish: 89%.
The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI).
Specifications
| STS | |
| Polyclonal | |
| Unconjugated | |
| Sts | |
| ArsC; ARSC1; Arylsulfatase C; ASC; ES; Estrone sulfatase; LOC402795 protein; LOW QUALITY PROTEIN: steryl-sulfatase; steryl-sulfatase; si:ch211-271l9.1; SSDD; Steroid sulfatase; steroid sulfatase (microsomal), arylsulfatase C, isozyme S; steroid sulfatase (microsomal), isozyme S; steroid sulphatase; steryl-sulfatase; Steryl-sulfate sulfohydrolase; STS; XLI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 412 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P08842 | |
| Sts | |
| Synthetic peptide directed towards the middle region of human STS. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction