Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110662 Supplier Invitrogen™ Supplier No. PA5110662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YEIIQQPMSY DNLTQRLTVE AAQFIQRNTE TPFLLVLSYL HVHTALFSSK DFAGKSQHGV YGDAVEEMDW SVGQILNLLD ELRLANDTLI YFTSDQGAHV EEVSSKGEIH GGSNGIY Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 40%.

The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sts
Gene Accession No. P08842
Gene Alias ArsC; ARSC1; Arylsulfatase C; ASC; ES; Estrone sulfatase; LOC402795 protein; LOW QUALITY PROTEIN: steryl-sulfatase; steryl-sulfatase; si:ch211-271l9.1; SSDD; Steroid sulfatase; steroid sulfatase (microsomal), arylsulfatase C, isozyme S; steroid sulfatase (microsomal), isozyme S; steroid sulphatase; steryl-sulfatase; Steryl-sulfate sulfohydrolase; STS; XLI
Gene Symbols Sts
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human STS. Recombinant protein control fragment (Product #RP-91919).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.