Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596591 Supplier Invitrogen™ Supplier No. PA596591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQYGHPSPL GMAAREELYS KVTPRRNRQQ RPGTIKHGSA LDVLLSMGFP RARAQKALAS TGGRSVQAAC DWLFSHVGDP FLDDPLPREY VLYLRPTGPL AQKLSDFWQQ SKQICGKNKA HNIFPHITLC QFFMCEDSKV DALGEALQTT VSRWKCKFSA PLPLELYTSS NFIGLFVKED SAEVLKKFAA DFAAEAASKT EVHVEPHKKQ LHVTLAYHFQ ASHLPTLEKL AQNIDVKLGC DWVATIFSRD IRFANHETLQ VIYPYTPQND DELELVPGDF IFMSPMEQTS TSEGWIYGTS; Positive Samples: SW620, SKOV3, Mouse brain, Mouse spleen, Mouse lung, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Sts1 encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBASH3B
Gene Accession No. Q8BGG7, Q8TF42
Gene Alias 2810457I06Rik; BB125008; Cbl-interacting protein p70; Cbl-interacting protein Sts-1; KIAA1959; MGC15437; nm23-phosphorylated unknown substrate; p70; RGD1310357; SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling 1, nm23-phosphorylated unknown substrate; STS1; STS-1; suppressor of T-cell receptor signaling 1; T-cell ubiquitin ligand 2; TULA2; TULA-2; tyrosine-protein phosphatase STS1/TULA2; UBASH3B; ubiquitin associated and SH3 domain containing B; ubiquitin associated and SH3 domain containing, B; ubiquitin-associated and SH3 domain-containing protein B
Gene Symbols UBASH3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human UBASH3B (NP_1162622).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315579, 72828, 84959
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.