Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STT3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100342 Supplier Invitrogen™ Supplier No. PA5100342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total STT3A.

This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STT3A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STT3A
Gene Accession No. P46977, P46978
Gene Alias AA408947; B5; BB081708; dolichyl-diphosphooligosaccharide protein glycotransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit STT3A; Integral membrane protein 1; integral transmembrane protein 1; intergral membrane protein 1; Itm1; MGC9042; Oligosaccharyl transferase subunit STT3A; RGD1565793; STT3, subunit of the oligosaccharyltransferase complex, homolog A; STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae); STT3A; STT3-A; STT3A, catalytic subunit of the oligosaccharyltransferase complex; STT3A, cataylic subunit of the oligosaccharyltransferase complex; STT3A, subunit of the oligosaccharyltransferase complex (catalytic); TMC; transmembrane conserved; transmembrane protein TMC
Gene Symbols STT3A
Host Species Rabbit
Immunogen A synthesized peptide derived from human STT3A(Accession P46977), corresponding to amino acid residues V422-T472.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16430, 3703, 500972
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.