Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STUB1 Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA530912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530912 Supplier Invitrogen™ Supplier No. MA530912
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human STUB1.

A recently identified protein, termed carboxyl terminus of hsc70-interacting protein (CHIP), has been shown to interact both with the constitutive form of hsc70 and the stress inducible form, hsp70. This novel 35 kDa cytoplasmic protein has been shown to be highly expressed in striated muscle in vivo. Additional studies have shown that this protein is expressed over a broad range of cultured tissues. Through immunoprecipitation experiments, CHIP has been shown to directly bind to the carboxyl terminus of hsc70 and hsp70 where it decreases ATPase activity and reduces overall chaperone efficiency. CHIP has also been identified as an important protein in the ubiquitin-proteasome system. CHIP contains a U-box domain and acts as an E3 ubiquitin-ligase in conjunction with hsc70 and hsp90.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STUB1
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene STUB1
Gene Accession No. Q9UNE7
Gene Alias 0610033N24Rik; 2210017D18Rik; 2310040B03Rik; Antigen NY-CO-7; AW046544; Carboxy terminus of Hsp70-interacting protein; CHIP; CLL-associated antigen KW-8; E3 ubiquitin-protein ligase CHIP; heat shock protein A binding protein 2 (c-terminal); HSPABP2; LA16c-313D11.6; NY-CO-7; PP1131; RING-type E3 ubiquitin transferase CHIP; SCAR16; SDCCAG7; serologically defined colon cancer antigen 7; STIP1 homology and U box-containing protein 1; STIP1 homology and U-box containing protein 1; STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase; Stub1; UBOX1
Gene Symbols STUB1
Host Species Mouse
Immunogen Recombinant Human STUB1 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.