Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Student Bacteria Experiment Kit
Shop All Fischer Technical Company Products

Click to view available options
Class Size:
24 Students (4 Groups)
Description
Includes
- Two 200mL bottles of prepared nutrient agar
- 20 radiation-sterilized Petri dishes
- 10 sterile swabs
- Four lab instruction sheets
Melt the media in a microwave, pour it into media plates and test for growth in four popular experiments.
Experiments:
- Test-common use items such as bathroom handles, doorknobs and water fountains
- Clean hand/dirty hand test: How many bacteria does soap eliminate?
- Clean mouth/dirty mouth test: How many bacteria does mouthwash eliminate?
- Which mouth contains more bacteria: a human's or a dog's?
Order Info
Class size: Four groups of five to six students.
Specifications
Specifications
Product Type | Student Bacteria Experiment Kit |
Grade | 4 to 12 |
For Use With (Application) | Allows groups of students to culture their own bacteria without an incubator |
Includes | Two 200ml bottles of prepared nutrient agar, 20 radiation-sterilized petri dishes, 10 sterile swabs, four lab instruction sheets |
Class Size | 24 Students (4 Groups) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction