Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Student Bacteria Science Kit
Shop All Aldon Products

Click to view available options
Class Size:
1 Student
Description
This Student Bacteria Science kit contains materials for students to use in investigating bacteria in their environment while learning proper microbiological techniques.
Students will test hands, mouth and classroom for bacteria, before and after cleaning.
Kit Includes:
- 2 x 200 ml Nutrient Agar
- 20 Sterile Petri Dishes
- 20 Sterile Cotton Swabs
Specifications
Specifications
Certifications/Compliance | Product SDSs |
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | Science Education |
Includes | 2 x 200 mL Nutrient Agar, 20 Sterile Petri Dishes, 20 Sterile Cotton Swabs |
Quantity | 1 |
Weight (English) | 2.5 lb. |
Dimensions (L x W x H) | 13 x 10 x 7 in. |
Class Size | 1 Student |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction