Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Manufacturing Student Thermometer
SCI_ED

Catalog No. S95600 Shop All Si Metric Manufacturing Limited Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +110°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S95600 -50°C to +110°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S95600 Supplier SI Manufacturing Supplier No. 95570

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

For all grade levels.

  • Red alcohol thermometer is recessed in molded plastic to prevent breakage
  • Can be immersed in hot liquids without danger of burning students' hands
  • Reads from -50° to +110°C (-58° to +230°F)

Order Info

Length: 5 in. (13cm)

Specifications

Length (English) Overall 5 in.
Length (Metric) Overall 127 mm
Material Plastic
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +110°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.