Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STUM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26897925UL
Description
STUM Polyclonal antibody specifically detects STUM in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
STUM | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
chromosome 1 open reading frame 95, hypothetical protein LOC375057, RP11-9C4.1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FVSAFTVLCGARTDLPDRHVCCVF | |
25 μL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
375057 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction