Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sturdy Magnetic Storage Pockets
SCI_ED

Catalog No. s04667
Click to view available options
Color:
Green, Blue, Purple, Red
Choking hazard. Small magnets.
Choking hazard. Small magnets.

Get students organized with sturdy magnetic storage pockets.

  • Hold 0.75 lbs.
  • Set of four
  • Four colors: green, blue, purple, red
  • L x W: 9.5 x 5.5 in. (24.13 x 13.8cm)

Specifications

Product Type Magnetic Pockets
Color Green, Blue, Purple, Red
Mounting Type Magnetic
Includes Velcro fastening lid
For Use With (Application) To help with classroom organization
Capacity (Metric) 350 g
Height (English) 6 in.
Height (Metric) 14 cm
Length (English) 9 in.
Length (Metric) 23 cm
Weight (English) 4 oz.
Weight (Metric) 0.113 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.