Learn More
Description
Includes
Stx-CLPesticides column, Stx-CLPesticides2 column, Universal Angled Y; Press-Tight connector and a Siltek-deactivated guard column
Requires
- Concentration in Solvent: 0.100g/dL each in water
- Volume: 1mL/ampule
- Shelf Life: 6 months
Compounds
- Acetaldehyde (75-07-0)
- Acetone (67-64-1)
- Acetonitrile (75-05-8)
- Ethanol (64-17-5)
- Ethyl acetate (141-78-6)
- 2-Propanol (Isopropanol) (67-63-0)
- Methanol (67-56-1)
- Methyl ethyl ketone (78-93-3)
- Baseline separations in less than 10 minutes
- Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
- Highly inert
- Stable to 330°C
Note:Columns are not preconnected in this kit.
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
| For Use With (Application) | Organochlorine Pesticides Analysis, Herbicides Analysis |
| Film Thickness | Not Applicable |
| Length (Metric) | 30 m |
| Polarity | Mid Polarity |
| Includes | Rtx™-CLPesticides column, Rtx™-CLPesticides2 column, deactivated universal angled Y Press-Tight™ connector, guard column |
| USP Type | None |
| Stationary Phase | Stx™ -CLPesticides |
| Type | Capillary Column Kit |
| Diameter (Metric) Inner | 0.25 mm |
| Max. Temperature (Metric) | 330°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
