Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589467 Supplier Invitrogen™ Supplier No. PA589467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKPAAQKSPS DLLDASAVSA TSRYIEEQQA TQQLIMDEQD QQLEMVSGSI QVLKHMSGRV GEELDEQGIM LDAFAQEMDH TQSRMDGVLR; Positive Samples: HeLa; Cellular Location: Golgi apparatus membrane, Single-pass type IV membrane protein.

SNARE involved in vesicular transport from the late endosomes to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene STX10
Gene Accession No. O60499
Gene Alias hsyn10; STX10; Syn10; syntaxin 10; syntaxin-10
Gene Symbols STX10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 76-165 of human STX10 (NP_0012585401).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8677
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.