Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STX11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106321
Description
Antibody detects endogenous levels of total Syntaxin 11.
This gene encodes a member of the syntaxin family. Syntaxins have been implicated in the targeting and fusion of intracellular transport vesicles. This family member may regulate protein transport among late endosomes and the trans-Golgi network. Mutations in this gene have been associated with familial hemophagocytic lymphohistiocytosis.
Specifications
| STX11 | |
| Polyclonal | |
| Unconjugated | |
| STX11 | |
| 5830405C08Rik; FHL4; HLH4; HPLH4; STX11; syntaxin 11; syntaxin-11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 292483, 74732, 8676 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75558, Q9D3G5 | |
| STX11 | |
| A synthesized peptide derived from human STX11(Accession O75558), corresponding to amino acid residues I159-S209. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction