Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STX12 Monoclonal Antibody (4E212)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA126001
Description
MA1-26001 detects a band of ~38 kDa corresponding to recombinant rat syntaxin 13. Suggested positive controls include rat or mouse brain extract.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
Specifications
STX12 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide | |
G3V7P1, Q9ER00 | |
STX12 | |
Recombinant fragment corresponding to the cytoplasmic domain of rat Syntaxin 13. | |
50 μg | |
Primary | |
Mouse, Rat, Hamster | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
'4E212 | |
Unconjugated | |
STX12 | |
AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13 | |
Mouse | |
Protein G | |
RUO | |
100226, 100773207, 65033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction