Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559407 Supplier Invitrogen™ Supplier No. PA559407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSYGPLDVYR NPGPSGPQLR DFSSIIQTCS GNIQRISQAT AQIKNLMSQL GTKQDSSKLQ ENLQQLQHST NQLAKETNEL LKE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX12
Gene Accession No. G3V7P1, Q86Y82, Q9ER00
Gene Alias AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13
Gene Symbols STX12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX12. Recombinant protein control fragment (Product #RP-97617).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100226, 23673, 65033
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.