Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117123 Supplier Invitrogen™ Supplier No. PA5117123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse kidney, Mouse brain, Rat kidney Immunogen sequence: MSYGPLDMYR NPGPSGPQLR DFSSIIQTCS GNIQRISQAT AQIKNLMSQL GTKQDSSKLQ ENLQQLQHST NQLAKETNEL LKELGSLPLP LSTSEQRQQR LQKERLMNDF SAALNNFQAV QRRVSEKEKE SIARARAGSR LSAEERQREE QLVSFDSHEE WNQMQSQEDE VAITEQDLEL IKERETAIRQ LEADILDVNQ IFKDLAMMIH DQGDLIDSIE ANVESSEVHV ERATEQLQRA AYYQKKSRKK MCILVLVLSV IILILGLIIW LVYKTK.

SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene STX12
Gene Accession No. G3V7P1, Q86Y82, Q9ER00
Gene Alias AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13
Gene Symbols STX12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human STX12 (NP_8031731).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100226, 23673, 65033
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.