Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STX17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540127
Description
Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Predicted reactivity: Mouse (89%), Rat (86%), Rhesus Monkey (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SNAREs, soluble N-ethylmaleimide-sensitive factor-attachment protein receptors, are essential proteins for fusion of cellular membranes. SNAREs localized on opposing membranes assemble to form a trans-SNARE complex, an extended, parallel four alpha-helical bundle that drives membrane fusion. STX17 is a SNARE of the autophagosome involved in autophagy through the direct control of autophagosome membrane fusion with the lysosome membrane. STX17 may also play a role in the early secretory pathway where it may maintain the architecture of the endoplasmic reticulum-Golgi intermediate compartment/ERGIC and Golgi and/or regulate transport between the endoplasmic reticulum, the ERGIC and the Golgi. (Uniprot).
Specifications
STX17 | |
Polyclonal | |
Unconjugated | |
STX17 | |
4833418L03Rik; 6330411F21Rik; 9030425C21Rik; AW048351; Stx17; syntaxin 17; syntaxin-17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
252853, 55014, 67727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.94 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P56962, Q9D0I4, Q9Z158 | |
STX17 | |
Recombinant protein encompassing a sequence within the center region of human STX17. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction