Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551535 Supplier Invitrogen™ Supplier No. PA551535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLEPAIQKFI KIVIPTDLER LRKHQINIEK YQRCRIWDKL HEEHINAGRT VQQLRSNIRE IEKLCLKVRK DDLVLLKRMI DPVKEEASAA TAEFLQLHLE SVEELKKQFN DEETLLQPPL TRSMTVGGAF HTTEAEASSQ SLTQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

SNAREs, soluble N-ethylmaleimide-sensitive factor-attachment protein receptors, are essential proteins for fusion of cellular membranes. SNAREs localized on opposing membranes assemble to form a trans-SNARE complex, an extended, parallel four alpha-helical bundle that drives membrane fusion. STX17 is a SNARE of the autophagosome involved in autophagy through the direct control of autophagosome membrane fusion with the lysosome membrane. STX17 may also play a role in the early secretory pathway where it may maintain the architecture of the endoplasmic reticulum-Golgi intermediate compartment/ERGIC and Golgi and/or regulate transport between the endoplasmic reticulum, the ERGIC and the Golgi. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX17
Gene Accession No. P56962
Gene Alias 4833418L03Rik; 6330411F21Rik; 9030425C21Rik; AW048351; Stx17; syntaxin 17; syntaxin-17
Gene Symbols STX17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX17. Recombinant protein control fragment (Product #RP-96776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.