Learn More
STX18 Mouse anti-Human, Clone: OTI2B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809423

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.Specifications
STX18 | |
Monoclonal | |
Unconjugated | |
STX18 | |
Ufe1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53407 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9P2W9 | |
STX18 | |
Human recombit protein fragment corresponding to amino acids 1-285 of human STX18 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.