Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551797 Supplier Invitrogen™ Supplier No. PA551797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKRVVDKKRL SKLEPEPNTK TRESTSSEKV SQSPSKDSEE NPATEERPEK ILAETQPELG TWGDGKGEDE LSPEEIQMFE QENQRLIGEM NSLFDEVRQI EGRVVEISRL QEIFTEKVLQ QEAEIDSIHQ LVVGATENIK EGNEDIREAI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX18
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX18
Gene Accession No. Q9P2W9
Gene Alias 1810035L21Rik; 4933425D03Rik; AU041562; cell growth-inhibiting gene 9 protein; GIG9; STX18; syntaxin 18; syntaxin-18; Ufe1
Gene Symbols STX18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX18. Recombinant protein control fragment (Product #RP-95933).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.