Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556519 Supplier Invitrogen™ Supplier No. PA556519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEIELSRDSH VSTTETEEQG VFLQQAVIYE REPVAERHLH EIQKLQESIN NLADNVQKFG QQQKSLVASM RRFSLLKRES TITKEIKIQA EYINRSLNDL VKEVKKSEVE NGPSSVVTRI LKSQHAAMFR HFQQIMFIYN D Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 30%.

Syntaxin 19, also known as STX19, is a 294 amino acid peripheral membrane protein that contains one t-SNARE coiled-coil homology domain and belongs to the syntaxin family, suggesting a role in synaptic vesicle fusion. The gene encoding Syntaxin 19 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX19
Gene Accession No. Q8N4C7
Gene Alias A030009B12Rik; Stx19; Syn9; syntaxin 19; syntaxin 9; syntaxin-19; Syntaxin-9
Gene Symbols STX19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX19. Recombinant protein control fragment (Product #RP-95940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 415117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.