Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STX1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116376
Description
STX1B Polyclonal Antibody for Western Blot
Potentially involved in docking of synaptic vesicles at presynaptic active zones. May mediate Ca(2+)-regulation of exocytosis acrosomal reaction in sperm.
Specifications
| STX1B | |
| Polyclonal | |
| Unconjugated | |
| STX1B | |
| GEFSP9; STX1B; STX1B1; STX1B2; syntaxin 1B; syntaxin-1B; Syntaxin-1B1; syntaxin-1B2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 112755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P61266 | |
| STX1B | |
| KLH conjugated synthetic peptide between 150-183 amino acids from the Central region of human STX1B. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction