Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STX7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581945
Description
Immunogen sequence: GVGGDPAQLA QRISSNIQKI TQCSVEIQRT LNQLGTPQDS PELRQQLQQK QQYTNQLAKE TDKYIKEFGS LPTTPSEQRQ RKIQKDRLVA EFTTSLTNFQ KVQRQAAERE KEFVARVRAS SRVSGSFPED SSKERNLVSW ESQTQPQV.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
Specifications
STX7 | |
Polyclonal | |
Unconjugated | |
Stx7 | |
AI315064; AI317144; AW107239; STX7; Syn7; syntaxin 7; Syntaxin7; syntaxin-7; syntaxin-7-like; syntaxin-7-like protein; Unknown (protein for MGC:128117) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53331, 60466, 8417 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15400, O70257, O70439 | |
Stx7 | |
Recombinant protein corresponding to Human STX7. Recombinant protein control fragment (Product #RP-95935). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction