Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ STXBP2 Recombinant Protein

Catalog No. 89010837
Click to view available options
Quantity:
10 μg
25 μg

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH02869
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 6813
Molecular Weight (g/mol) 90.64
Name STXBP2 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MAPSGLKAVVGEKILSGVIRSVKKDGEWKVLIMDHPSMRILSSCCKMSDILAEGITIVEDINKRREPIPSLEAIYLLSPTEKALIKDFQGTPTFTYKAAHIFFTDTCPEPLFSELGRSRLAKVVKTLKEIHLAFLPYEAQVFSLDAPHSTYNLYCPFRAEERTRQLEVLAQQIATLCATLQEYPAIRYRKGPEDTAQLAHAVLAKLNAFKADTPSLGEGPEKTRSQLLIMDRAADPVSPLLHELTFQAMAYDLLDIEQDTYRYETTGLSEAREKAVLLDEDDDLWVELRHMHIADVSKKVTELLRTFCESKRLTTDKANIKDLSQILKKMPQYQKELNKYSTHLHLADDCMKHFKGSVEKLCSVEQDLAMGSDAEGEKIKDSMKLIVPVLLDAAVPAYDKIRVLLLYILLRNGVSEENLAKLIQHANVQAHSSLIRNLEQLGGTVTNPGGSGTSSRLEPRERMEPTYQLSRWTPVIKDVMEDAVEDRLDRNLWPFVSDPAPTASSQAAVSARFGHWHKNKAGIEARAGPRLIVYVMGGVAMSEMRAAYEVTRATEGKWEVLIGSSHILTPTRFLDDLKALDKKLEDIALP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias Hunc18b/MUNC18-2/UNC18-2/UNC18B/pp10122
Common Name STXBP2
Gene Symbol STXBP2
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.