Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STXBP3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537892
Description
This antibody is predicted to react with human, dog and pig based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Specifications
STXBP3 | |
Polyclonal | |
Unconjugated | |
STXBP3 | |
Mammalian homolog of Unc-18c; MUNC-18-3; MUNC18-3; MUNC18C; Munc-18c; Munc18-c; Platelet Sec1 protein; protein unc-18 homolog 3; Protein unc-18 homolog C; PSP; Stxbp3; Stxbp3a; Sxtbp3; syntaxin 4 binding protein; syntaxin binding protein 3; syntaxin binding protein 3A; syntaxin-binding protein 3; Syntaxin-BP3; unc-18 homolog 3; Unc18-3; Unc18c; unc-18C | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
114095, 20912 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q60770 | |
STXBP3 | |
Peptide with sequence SKSIRRCKEINIS, from the internal region of STXBP3 (aa 127-139). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction