Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STYK1 Monoclonal Antibody (2H2F10)
GREENER_CHOICE

Catalog No. PIMA515372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515372 Supplier Invitrogen™ Supplier No. MA515372
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15372 targets STYK1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15372 immunogen is purified recombinant fragment of STYK1 expressed in E. Coli..

STYK1, a probable tyrosine protein-kinase, which has strong transforming capabilities on a variety of cell lines. When overexpressed, it can also induce tumor cell invasion as well as metastasis in distant organs. May act by activating both MAP kinase and phosphatidylinositol 3'-kinases (PI3K) pathways. It is widely expressed; highly expressed in brain, placenta and prostate. STYK1 is expressed in tumor cells such as hepatoma cells LO2, cervix carcinoma cells HeLa, ovary cancer cells Ho8910 and chronic myelogenous leukemia cells K562, but not in other tumor cells such as epidermoid carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STYK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H2F10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene STYK1
Gene Accession No. Q6J9G0
Gene Alias 9130025L13; AI326477; DKFZp761P1010; mNOK; Nok; NOK kinase; novel oncogene with kinase domain; Protein PK-unique; RGD1564211; Serine/threonine/tyrosine kinase 1; Styk1; SuRTK106; tyrosine-protein kinase STYK1
Gene Symbols STYK1
Host Species Mouse
Immunogen Purified recombinant fragment of STYK1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.