Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STYK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583501 Supplier Invitrogen™ Supplier No. PA583501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGPQGIAPVP PPRDLSWEAG HGGNVALPLK ETSVENFLGA TTPALAKLQV PREQLSEVLE QICSGSCGPI FRANMNTGDP SKPKSVILKA LKEPAGLHEV QDFLGRIQFH QYLGKHKNLV QLEGCCTEKL PLYM Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 73%.

STYK1, a probable tyrosine protein-kinase, which has strong transforming capabilities on a variety of cell lines. When overexpressed, it can also induce tumor cell invasion as well as metastasis in distant organs. May act by activating both MAP kinase and phosphatidylinositol 3'-kinases (PI3K) pathways. It is widely expressed; highly expressed in brain, placenta and prostate. STYK1 is expressed in tumor cells such as hepatoma cells LO2, cervix carcinoma cells HeLa, ovary cancer cells Ho8910 and chronic myelogenous leukemia cells K562, but not in other tumor cells such as epidermoid carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STYK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STYK1
Gene Accession No. Q6J9G0
Gene Alias 9130025L13; AI326477; DKFZp761P1010; mNOK; Nok; NOK kinase; novel oncogene with kinase domain; Protein PK-unique; RGD1564211; Serine/threonine/tyrosine kinase 1; Styk1; SuRTK106; tyrosine-protein kinase STYK1
Gene Symbols STYK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STYK1. Recombinant protein control fragment (Product #RP-89965).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.