Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene, 99.5% stab. with 4-tert-butylcatechol

Catalog No. AA16199AK
Click to view available options
Quantity:
50 mL
250 mL
1 L

CAS: 100-42-5 | C8H8 | 104.15 g/mol

Styrene is used in the preparation of polystyrene, which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include in refrigerator liners, in medical devices and in small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN) which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31°C
Color Colorless
Density 0.906 g/mL
pH neutral
Boiling Point 145°C to 146°C
Flash Point 31°C (88°F)
Odor Pungent
Refractive Index 1.546
Quantity 250 mL
UN Number UN2055
Beilstein 1071236
Sensitivity Air and light sensitive
Merck Index 14,8860
Solubility Information Miscible with alcohols,ethers and carbon disulfide. Immiscible with water.
IUPAC Name styrene
Formula Weight 104.15
Percent Purity 99.5%
Grade Technical
Physical Form Liquid
Chemical Name or Material Styrene, Stabilized with 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372
Hazard Statement GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED
EINECSNumber 202-851-5
RTECSNumber WL3675000
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.