Alfa Aesar™ Styrene-d8, 98% (Isotopic) stab. with 4-tert-butylcatechol

Manufacturer: Alfa Aesar™ 4366706

Quantity 5g

 View more versions of this product

Catalog No. AA4366706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19361-62-7
    Molecular Formula C8D8
    Linear Formula C6D5CD=CD2
    Formula Weight 112.2
    MDL Number MFCD00044231
    UN Number UN2055
    Beilstein 2327045
    Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic

    Specifications

    Formula Weight 112.2
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula C6D5CD=CD2
    UN Number UN2055
    TSCA TSCA
    Density 0.979g/mL
    Boiling Point 50° to 51°C (25mmHg)
    Color Colorless
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED
    Flash Point 31°C (88°F)
    Refractive Index 1.5445
    MDL Number MFCD00044231
    Storage Note 1
    • Ampouled under argon
    Recommended Storage 2°C to 8°C
    Quantity 5g
    CAS 19361-62-7
    Beilstein 2327045
    Health Hazard 1 Warning
    Sensitivity Air Sensitive
    Molecular Formula C8D8
    Health Hazard 2 GHS H Statement
    H226-H332-H315-H319
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Physical Form Liquid
    Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic
    Assay 98%

    GHS H Statement
    H226-H332-H315-H319
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    2°C to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit