Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene oxide, 97+%, Acros Organics™

Click to view available options
Quantity:
10mL
250mL
1L
2.5L
Packaging:
Glass bottle
96-09-3
C8H8O
120.15
MFCD00005121
AWMVMTVKBNGEAK-UHFFFAOYSA-N
styrene oxide,phenyloxirane,1,2-epoxyethylbenzene,phenylethylene oxide,epoxystyrene,styrene epoxide,styryl oxide,oxirane, phenyl,1-phenyloxirane,phenethylene oxide
7276
CHEBI:17907
2-phenyloxirane
C1C(O1)C2=CC=CC=C2
96-09-3
C8H8O
120.15
MFCD00005121
AWMVMTVKBNGEAK-UHFFFAOYSA-N
styrene oxide,phenyloxirane,1,2-epoxyethylbenzene,phenylethylene oxide,epoxystyrene,styrene epoxide,styryl oxide,oxirane, phenyl,1-phenyloxirane,phenethylene oxide
7276
CHEBI:17907
2-phenyloxirane
C1C(O1)C2=CC=CC=C2
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5330 to 1.5360
Specific Gravity 1.05
Melting Point -37.0°C
Flash Point 74°C
Boiling Point 194.0°C
Density 1.0500g/mL
Quantity 10mL
Color Colorless to Yellow
Assay Percent Range 97+%
Beilstein 17, 49
Solubility Information Solubility in water: 1.5g/L (20°C). Other solubilities: soluble in hexane,petroleum ether,acetone,,alcohol,ether and benzene
Formula Weight 120.15
Physical Form Liquid
Percent Purity ≥97%
Chemical Name or Material Styrene oxide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic in contact with skin.
May cause cancer.

GHS P Statement:
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger