Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STYX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559134 Supplier Invitrogen™ Supplier No. PA559134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVKLEFPSLP QCKEDAEEWT YPMRREMQEI LPGLFLGPYS SAMKSKLPVL QKHGITHIIC IRQNIEANFI KPN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen STYX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STYX
Gene Accession No. Q8WUJ0
Gene Alias Inactive tyrosine-protein phosphatase STYX; Phosphoserine/threonine/tyrosine interaction protein; protein tyrosine phosphatase-like protein; serine/threonine/tyrosine interacting protein; serine/threonine/tyrosine-interacting protein; STYX
Gene Symbols STYX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STYX. Recombinant protein control fragment (Product #RP-97616).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6815
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.