Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STYXL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530426
Description
Recommended positive controls: A549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalytically inactive phosphatase (PubMed:20180778, PubMed:23163895). By binding to G3BP1, inhibits the formation of G3BP1-induced stress granules (PubMed:20180778, PubMed:23163895). Does not act by protecting the dephosphorylation of G3BP1 at 'Ser-149' (PubMed:23163895). Inhibits PTPMT1 phosphatase activity (PubMed:24709986). By inhibiting PTPMT1, positively regulates intrinsic apoptosis (PubMed:21262771). May play a role in the formation of neurites during neuronal development (PubMed:29250526). [UniProt].
Specifications
STYXL1 | |
Polyclonal | |
Unconjugated | |
STYXL1 | |
dual specificity phosphatase 24 (putative); dual specificity phosphatase inhibitor MK-STYX; Dual specificity protein phosphatase 24; DUSP24; Inactive dual specificity protein phosphatase MK-STYX; Map kinase phosphatase-like protein MK-STYX; MKSTYX; MK-STYX; serine/threonine/tyrosine interacting-like 1; serine/threonine/tyrosine-interacting-like protein 1; STYXL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y6J8 | |
STYXL1 | |
Full length human STYXL1 Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction