Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SU 16f
GREENER_CHOICESDP

Catalog No. 330450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-045-0 50 mg
33-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-045-0 Supplier R&D Systems™ Supplier No. 3304/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective PDGFRβ inhibitor

Potent and selective platelet-derived growth factor receptor β (PDGFRβ) inhibitor (IC50 = 10 nM) that displays > 14-fold, > 229-fold and > 10000-fold selectivity over VEGFR2, FGFR1 and EGFR respectively. Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 = 0.11 μM).

Chemical Identifiers

CAS 251356-45-3
Molecular Formula C24H22N2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 386.44
Purity >98%
Molecular Weight (g/mol) 386.451
Chemical Name or Material SU 16f
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.