Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SU 5416
GREENER_CHOICE

Catalog No. 303710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-371-0 10 mg
30-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-371-0 Supplier Tocris Bioscience™ Supplier No. 3037/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

VEGFR inhibitor. Also inhibits KIT, RET, MET and FLT3

Inhibitor of vascular endothelial growth factor receptor (VEGFR) that also inhibits other tyrosine kinases KIT, MET, FLT3 and RET. Displays no activity against EGFR, HER2, IGF1R and PDGFR. Inhibits tumor vascularization and growth of multiple tumor types.

Chemical Identifiers

CAS 204005-46-9
Molecular Formula C15H14N2O

Specifications

Quantity 10 mg
Formula Weight 238.28
Purity >99%
Molecular Weight (g/mol) 238.29
Chemical Name or Material SU 5416
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.