Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUB1 Antibody (04), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306541AF488
Description
SUB1 Monoclonal antibody specifically detects SUB1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SUB1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
04 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
activated RNA polymerase II transcription cofactor 4, activated RNA polymerase II transcriptional coactivator p15, MGC102747, p15, PC4p14P15, Positive cofactor 4, RPO2TC1, SUB1 homolog, SUB1 homolog (S. cerevisiae) | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SUB1 (P53999; Met1-Leu127). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
10923 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction