Learn More
Thermo Scientific™ Subcellular Protein Fractionation Kit for Cultured Cells PROMO
Description
The kit enables the stepwise separation and extraction of cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal proteins from mammalian cultured cells. Extracts obtained with the Subcellular Protein Fractionation Kit for Cultured Cells are compatible with a variety of downstream applications including Western blotting, protein assays, electrophoretic mobility shift assays and reporter-gene and enzyme-activity assays. Fractionation of subcellular proteins enables protein localization assessment and protein enrichment from specific cellular compartments. The Thermo Scientific Subcellular Protein Fractionation Kit includes a combination of reagents for stepwise lysis of cells into functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours. Extracts from each subcellular compartment generally have less than 15% contamination between fractions, which is sufficient purity for most experiments studying protein localization and redistribution.
Highlights:
Concise – extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than three hours
Convenient – perform a simple procedure without using ultracentrifugation over gradients
Complete – obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
Cultured Cells – developed and optimized for use with cultured mammalian cells; for tissue samples use the Subcellular Fractionation Kit for Tissues, Part No. 87790
Compatible – use with a variety of downstream applications including Western blotting, protein assays, electrophoretic mobility shift assays and reporter-gene and enzyme-activity assays
Small-scale – kit provides sufficient reagents to fractionate 50 cell pellets of 20μL each, equivalent to a total of 2g of cell paste or tissue
Includes:
Four extraction buffers, micrococcal nuclease enzyme, calcium chloride solution and protease inhibitor cocktail
Recommended for:
Extraction and enrichment of proteins from different cellular compartments; Separation and profiling of protein from the cytoplasm, nucleus, membrane and cytoskeleton from the same cell sample; Studying protein translocation
Specifications
Specifications
| Product Type | Subcellular Protein Fractionation Kit |
| Content And Storage | Upon receipt store Kit at -20°C, or store individual components as indicated above. Kit is shipped with dry ice. |
| Form | Liquid |
| Buffer | Lysis Buffer |
| Reagent Type | Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation |
| Sufficient For | 50 Samples |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.