Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suberic acid, 99%

Catalog No. p-194051
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 505-48-6 | C8H14O4 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-48-6
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00004428
InChI Key TYFQFVWCELRYAO-UHFFFAOYSA-N
Synonym suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
PubChem CID 10457
ChEBI CHEBI:9300
IUPAC Name octanedioic acid
SMILES OC(=O)CCCCCCC(O)=O

Specifications

Melting Point 141°C to 144°C
Color White to Cream
Boiling Point 230°C (15.0 mmHg)
Flash Point 203°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HO2C(CH2)6CO2H
Quantity 100 g
Beilstein 02, IV, 2028
Merck Index 15, 8992
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: 1g/172mL ether,soluble in alcohol,practically insoluble in chloroform,soluble in polar solvents
Viscosity 6.2 mPa.s (150°C)
Formula Weight 174.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Suberic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-010-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.