Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Suberonitrile 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
250 g
Specifications
Specifications
Color | Colorless |
Boiling Point | 172°C |
Quantity | 250 g |
UN Number | 3276 |
Formula Weight | 136.20 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Suberonitrile |
Safety and Handling
EINECSNumber | (2)-3619 |
RTECSNumber | WM2131000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction