Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Substance P

Catalog No. p-5384969
Click to view available options
Quantity:
5 mg
25 mg

For cellular and molecular biology applications | 1347.64 g/mol

Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Format Powder
For Use With (Application) A neurokinin NK-1 receptor agonist
Molecular Weight (g/mol) 1347.64
Product Type Substance P
Quantity 25 mg
Regulatory Status RUO - research use only
Sequence Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.