Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Substance P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575165 Supplier Invitrogen™ Supplier No. PA575165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).

Substance P (SP) is currently thought to be one of the most important neurotransmitters and neurotransmitters in the brain. It is an undecapeptide belonging to the tachykinin family, which consists of excitatory neuropeptides synthesised in neuronal and glial cells of the human central and peripheral nervous system. Substance P, Neurokinin A, Neuropeptide K, and Neuropeptide gamma are all generated by post-translation cleavage of the precursor Protachykinin-1. Substance P forms the major endogenous ligand for Neurokinin 1 Receptor. The pharmacology of Substance P has been associated with a number of neurological and psychiatric disorders, namely nociception, migraine, asthma, nausea, inflammatory bowel syndrome, urinary incontinence, anxiety and depression. It has also been linked to obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Substance P
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TAC1
Gene Accession No. P06767, P20366, P41539
Gene Alias 4930528L02Rik; C-terminal-flanking peptide; Hs.2563; MGC26719; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK1; NK-1; NK2; Nka; Nkna; NPK; PPT; Ppt5fl; PPTA; PPT-A; PPTA3; precursor 1; preprotachykinin; preprotachykinin A; protachykinin; protachykinin-1; RAT TAC; RATPPTA3; SP; substance K; Substance P; TAC; Tac1; TAC2; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Symbols TAC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to mature of Substance P of human origin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21333, 24806, 6863
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.