Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Substrate Reagent Pack
GSA_VASDP

Catalog No. DY999 Shop All Novus Products
Change view
Click to view available options
Quantity:
16 x 12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY999 16 x 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY999 Supplier R&D Systems™ Supplier No. DY999
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

8 vials Color A, 8 vials Color B Product in process of being discontinued and replaced by new Next generation pack

Warm to room temperature before use. Color Reagent A and Color Reagent B should be mixed together in equal volumes within 15 minutes of use. Protect from light. Add 100μL of the mixture to each well and incubate at room temperature for 20 minutes while protecting from light. Add Stop Solution (Catalog # DY994) and read the microplate.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 16 x 12 mL
For Use With (Application) ELISA
Product Line R&D Systems
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.