Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Succinic Acid
SCI_ED

Catalog No. s25790 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Crystals

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Linear Formula (CH2COOH)2
Formula Weight 118.09
Density 1.56g/mL
Quantity 100 g
Packaging Crystals
Chemical Name or Material Succinic Acid
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.