Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid ACS AR Granular, Macron Fine Chemicals™

Catalog No. p-3178358
Click to view available options
Quantity:
500 g
Packaging:
POLYSTORMOR

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Formula Weight 118.09
Grade ACS
Quantity 500 g
Packaging POLYSTORMOR
Physical Form Granular
Chemical Name or Material Succinic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.