Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid (Crystalline/Certified), Fisher Chemical

Catalog No. a294500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

pH 2.7
Melting Point 185°C
Boiling Point 235°C
Color White
Assay Percent Range 99.0 to 100.3 %
Linear Formula HOOCCH2CH2COOH
Formula Weight 118.09
Percent Purity 99.0 to 100.3%
Grade Certified
Density 1.56g/cm³
Quantity 500 g
Flash Point 206°C
Ignition Residue 0.020% max.
Packaging Glass Bottle
Physical Form Powder/Solid
Vapor Pressure 0.0022mmHg at 19°C
Chemical Name or Material Succinic Acid
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.