Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic acid, disodium salt, 99%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
150-90-3
C4H4Na2O4
162.052
MFCD00002790
ZDQYSKICYIVCPN-UHFFFAOYSA-L
disodium succinate,sodium succinate,disodium butanedioate,butanedioic acid, disodium salt,soduxin,succinic acid disodium salt,jantaran sodny czech,unii-v8zgc8isr3,succinic acid, disodium salt,fema no. 3277
9020
CHEBI:63675
disodium;butanedioate
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
150-90-3
C4H4Na2O4
162.052
MFCD00002790
ZDQYSKICYIVCPN-UHFFFAOYSA-L
disodium succinate,sodium succinate,disodium butanedioate,butanedioic acid, disodium salt,soduxin,succinic acid disodium salt,jantaran sodny czech,unii-v8zgc8isr3,succinic acid, disodium salt,fema no. 3277
9020
CHEBI:63675
disodium;butanedioate
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
Assay Percent Range 99%
Linear Formula NaO2CCH2CH2CO2Na
Beilstein 02, 606
Merck Index 15, 8808
Formula Weight 162.05
Physical Form Crystalline Powder
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 0.3% max. (120°C)
Packaging Plastic bottle
Solubility Solubility in water: 300g/L (25°C).
Melting Point >300.0°C
Color White
Quantity 100g
Chemical Name or Material Succinic acid, disodium salt, 99%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 205-778-7

RTECSNumber : WM7751000

TSCA : TSCA